2-Bromo-4-chloro-5-iodoanisole ≥95%

Supplier: AOBChem USA

Warning

18674-5G 18674-1G 18674-25G 18674-500MG
77631-880EA 970 USD
77631-880 77637-032 77657-802 77645-020
2-Bromo-4-chloro-5-iodoanisole ≥95%
2-Bromo-4-chloro-5-iodoanisole

Precaución: For Research Use Only

Formula: C₇H₅BrClIO
Storage Temperature: Ambient
MDL Number: MFCD11845921
CAS Number: 1160574-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR