3-Ethylbenzene-1,2-diol ≥97%

Supplier: AOBChem USA

Warning

186499-5G 186499-25G 186499-1G 186499-500MG
77634-244EA 359 USD
77634-244 77657-800 77641-026 77649-074
3-Ethylbenzene-1,2-diol ≥97%
3-Ethylbenzene-1,2-diol

Precaución: For Research Use Only

Formula: C₈H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD21604516
CAS Number: 933-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR