4-(Benzyloxy)-2-bromophenol ≥95%

Supplier: AOBChem USA

Warning

18567-5G 18567-1G 18567-500MG
77627-970EA 1484.23 USD
77627-970 77637-028 77652-434
4-(Benzyloxy)-2-bromophenol ≥95%
4-(Benzyloxy)-2-bromophenol

Precaución: For Research Use Only

Formula: C₁₃H₁₁BrO₂
MW: 279.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03926352
CAS Number: 79352-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR