3,5-Dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

18468-5G 18468-25G 18468-1G 18468-500MG
77634-236EA 487 USD
77634-236 77653-976 77641-016 77649-060
3,5-Dimethyl-1,1'-biphenyl ≥95%
3,5-Dimethyl-1,1'-biphenyl

Precaución: For Research Use Only

Formula: C₁₄H₁₄
Storage Temperature: Ambient
MDL Number: MFCD09031096
CAS Number: 17057-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR