3,5-Dimethylbiphenyl-4-ylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18467-1G 18467-5G 18467-500MG
77637-026EA 514 USD
77637-026 77657-792 77645-014
3,5-Dimethylbiphenyl-4-ylboronic acid ≥97%
3,5-Dimethylbiphenyl-4-ylboronic acid

Precaución: For Research Use Only

Formula: 226.079 g/mol
MW: C₁₄H₁₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD26045241
CAS Number: 1057654-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR