(3-Chloro-2-fluorophenyl)(cyclohexyl)methanol ≥97%

Supplier: AOBChem USA

Warning

184450-5G 184450-25G 184450-1G 184450-500MG
77631-874EA 1381 USD
77631-874 77653-974 77641-014 77645-012
(3-Chloro-2-fluorophenyl)(cyclohexyl)methanol ≥97%
(3-Chloro-2-fluorophenyl)(cyclohexyl)methanol

Precaución: For Research Use Only

Formula: C₁₃H₁₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD20400991
CAS Number: 1339038-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR