Cyclohexyl(4-fluoro-2-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

184444-5G 184444-25G 184444-1G 184444-500MG
77627-968EA 1484.23 USD
77627-968 77635-494 77641-012 77649-058
Cyclohexyl(4-fluoro-2-methylphenyl)methanol ≥95%
Cyclohexyl(4-fluoro-2-methylphenyl)methanol

Precaución: For Research Use Only

Formula: C₁₄H₁₉FO
Storage Temperature: Ambient
MDL Number: MFCD17222664
CAS Number: 1343179-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR