2-Bromo-5-methyl-4-pyridinecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

183875-5G 183875-1G 183875-500MG
77653-968EA 2674 USD
77653-968 77641-010 77649-052
2-Bromo-5-methyl-4-pyridinecarboxylic acid ≥95%
2-Bromo-5-methyl-4-pyridinecarboxylic acid

Precaución: For Research Use Only

Formula: 216.03 g/mol
MW: C₇H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD16610193
CAS Number: 1211535-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR