5-Chloro-3-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18376-5G 18376-1G 18376-25G 18376-500MG
77631-872EA 1206 USD
77631-872 77637-022 77653-966 77649-050
5-Chloro-3-fluoro-2-methoxybenzaldehyde ≥97%
5-Chloro-3-fluoro-2-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD22062900
CAS Number: 145742-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR