1-(2,5-Difluorophenyl)-2-cyclohexen-1-ol ≥95%

Supplier: AOBChem USA

Warning

183141-5G 183141-1G 183141-25G 183141-500MG
77631-870EA 1357 USD
77631-870 77637-020 77657-786 77649-048
1-(2,5-Difluorophenyl)-2-cyclohexen-1-ol ≥95%
1-(2,5-Difluorophenyl)-2-cyclohexen-1-ol

Precaución: For Research Use Only

Formula: 210.22 g/mol
MW: C₁₂H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD34857515
CAS Number: 1594998-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR