4-(3-Fluoro-2-methylphenyl)tetrahydro-2H-thiopyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

182906-5G 182906-1G 182906-500MG
77627-966EA 1599.59 USD
77627-966 77637-018 77649-046
4-(3-Fluoro-2-methylphenyl)tetrahydro-2H-thiopyran-4-ol ≥95%
4-(3-Fluoro-2-methylphenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For Research Use Only

Formula: 226.31 g/mol
MW: C₁₂H₁₅FOS
Storage Temperature: Ambient
MDL Number: MFCD31693975
CAS Number: 2254571-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR