(3,5-Diisopropylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

18286-5G 18286-1G 18286-500MG
77627-964EA 1484.23 USD
77627-964 77637-016 77649-044
(3,5-Diisopropylphenyl)boronic acid ≥95%
(3,5-Diisopropylphenyl)boronic acid

Precaución: For Research Use Only

Formula: 206.089 g/mol
MW: C₁₂H₁₉BO₂
Storage Temperature: Ambient
MDL Number: MFCD28147712
CAS Number: 263397-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR