Cyclohexyl(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

182397-5G 182397-25G
77634-228EA 136 USD
77634-228 77653-962
Cyclohexyl(phenyl)methanol ≥95%
Cyclohexyl(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00019357
CAS Number: 945-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR