3-Chloro-2-fluoro-α,α,5-trimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

181939-5G 181939-1G 181939-500MG
77653-960EA 2348 USD
77653-960 77641-006 77649-042
3-Chloro-2-fluoro-α,α,5-trimethylbenzenemethanol ≥95%
3-Chloro-2-fluoro-α,α,5-trimethylbenzenemethanol

Precaución: For Research Use Only

Formula: 202.65 g/mol
MW: C₁₀H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD28526647
CAS Number: 1780314-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR