Ethyl 3-chloro-2-fluoro-5-methyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

181928-5G 181928-1G 181928-500MG
77653-958EA 2314 USD
77653-958 77641-004 77649-040
Ethyl 3-chloro-2-fluoro-5-methyl-α-oxobenzeneacetate ≥95%
Ethyl 3-chloro-2-fluoro-5-methyl-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 244.65 g/mol
MW: C₁₁H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34857080
CAS Number: 1897094-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR