1,3-Dibromo-2-ethoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

18186-5G 18186-25G 18186-1G 18186-500MG
77634-226EA 325 USD
77634-226 77653-954 77641-002 77645-002
1,3-Dibromo-2-ethoxy-5-methylbenzene ≥97%
1,3-Dibromo-2-ethoxy-5-methylbenzene

Precaución: For Research Use Only

Formula: C₉H₁₀Br₂O
Storage Temperature: Ambient
MDL Number: MFCD18447594
CAS Number: 1266253-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR