2,2-Difluoro-4-(trimethylsilyl)-1,3-benzodioxole ≥95%

Supplier: AOBChem USA

Warning

181377-5G 181377-25G 181377-1G 181377-500MG
77627-962EA 1484.23 USD
77627-962 77653-950 77640-998 77649-032
2,2-Difluoro-4-(trimethylsilyl)-1,3-benzodioxole ≥95%
2,2-Difluoro-4-(trimethylsilyl)-1,3-benzodioxole

Precaución: For Research Use Only

Formula: 230.28 g/mol
MW: C₁₀H₁₂F₂O₂Si
Storage Temperature: Ambient
MDL Number: MFCD30178360
CAS Number: 126120-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR