2,2-Difluoro-5-methyl-1,3-benzodioxole ≥95%
Supplier: AOBChem USA
|
Warning
|
181364-5G
181364-1G
181364-500MG
77627-960EA
1667.57
USD
77627-960
77640-996
77649-030
2,2-Difluoro-5-methyl-1,3-benzodioxole ≥95%
2,2-Difluoro-5-methyl-1,3-benzodioxole
Precaución: For Research Use Only
Formula:
172.13 g/mol MW: C₈H₆F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18087715 CAS Number: 68119-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...