4-Bromo-2-(methylthio)benzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
18130-5G
18130-1G
18130-25G
18130-500MG
77631-864EA
1007
USD
77631-864
77637-008
77653-948
77644-998
4-Bromo-2-(methylthio)benzaldehyde ≥97%
4-Bromo-2-(methylthio)benzaldehyde
Precaución: For Research Use Only
Formula:
C₈H₇BrOS Storage Temperature: Ambient |
MDL Number:
MFCD18887984 CAS Number: 918350-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...