α,α-Diethyl-2,5-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180935-5G 180935-25G 180935-1G 180935-500MG
77631-862EA 1034 USD
77631-862 77657-778 77640-990 77644-994
α,α-Diethyl-2,5-difluorobenzenemethanol ≥95%
α,α-Diethyl-2,5-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₄F₂O
Storage Temperature: Ambient
MDL Number: MFCD17223838
CAS Number: 1340518-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR