3,5-Diiodo-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

180476-1G 180476-5G 180476-500MG
77634-222EA 941 USD
77634-222 77657-776 77649-026
3,5-Diiodo-4-methylbenzoic acid ≥97%
3,5-Diiodo-4-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₈H₆I₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27921059
CAS Number: 10388-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR