5-Bromo-3-chloro-2-methoxypyridine-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

17989-5G 17989-1G 17989-25G 17989-500MG
77631-858EA 1094 USD
77631-858 77637-000 77657-772 77644-990
5-Bromo-3-chloro-2-methoxypyridine-4-carboxaldehyde ≥97%
5-Bromo-3-chloro-2-methoxypyridine-4-carboxaldehyde

Precaución: For Research Use Only

Formula: 250.477 g/mol
MW: C₇H₅BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD13181627
CAS Number: 1224604-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR