1-[2-Fluoro-4-methyl-3-(methylthio)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

179720-5G 179720-1G 179720-25G 179720-500MG
77627-952EA 1484.23 USD
77627-952 77636-998 77657-770 77644-986
1-[2-Fluoro-4-methyl-3-(methylthio)phenyl]ethanone ≥95%
1-[2-Fluoro-4-methyl-3-(methylthio)phenyl]ethanone

Precaución: For Research Use Only

Formula: 198.26 g/mol
MW: C₁₀H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD34856285
CAS Number: 2091022-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR