2-(Methylsulfanyl)-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

179438-5G 179438-1G 179438-25G 179438-500MG
77631-856EA 842 USD
77631-856 77636-994 77657-768 77644-982
2-(Methylsulfanyl)-5-(trifluoromethyl)benzoic acid ≥95%
2-(Methylsulfanyl)-5-(trifluoromethyl)benzoic acid

Precaución: For Research Use Only

Formula: C₉H₇F₃O₂S
MDL Number: MFCD16853045
CAS Number: 53985-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR