2-Chloro-3-fluoro-5-picoline-4-boronic acid ≥97%

Supplier: AOBChem USA

Warning

17890-1G 17890-5G 17890-500MG
77634-218EA 1041 USD
77634-218 77653-932 77649-014
2-Chloro-3-fluoro-5-picoline-4-boronic acid ≥97%
2-Chloro-3-fluoro-5-picoline-4-boronic acid

Precaución: For Research Use Only

Formula: 189.38 g/mol
MW: C₆H₆BClFNO₂
Storage Temperature: Ambient
CAS Number: 1072952-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR