4-Chloro-2-(cyclopropylmethoxy)-1-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

178523-5G 178523-25G 178523-1G 178523-500MG
77627-948EA 1484.23 USD
77627-948 77635-490 77636-990 77644-972
4-Chloro-2-(cyclopropylmethoxy)-1-iodobenzene ≥95%
4-Chloro-2-(cyclopropylmethoxy)-1-iodobenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₀ClIO
Storage Temperature: Ambient
MDL Number: MFCD29092625
CAS Number: 1369848-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR