2-Bromo-5-methoxypyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

177417-1G 177417-5G 177417-500MG
77636-984EA 735 USD
77636-984 77653-924 77649-006
2-Bromo-5-methoxypyridin-3-ol ≥97%
2-Bromo-5-methoxypyridin-3-ol

Precaución: For Research Use Only

Formula: 204.021 g/mol
MW: C₆H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD15144624
CAS Number: 1256833-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR