1-Bromo-2,3-dichloro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

177392-5G 177392-500MG 177392-25G 177392-1G
77634-216EA 386 USD
77634-216 77652-428 77657-760 77640-978
1-Bromo-2,3-dichloro-5-iodobenzene ≥97%
1-Bromo-2,3-dichloro-5-iodobenzene

Precaución: For Research Use Only

Formula: 351.795 g/mol
MW: C₆H₂BrCl₂I
Storage Temperature: Ambient
MDL Number: MFCD30721706
CAS Number: 2307629-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR