1-(2-Fluoro-4-iodo-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

176985-5G 176985-25G 176985-1G 176985-500MG
77627-942EA 1484.23 USD
77627-942 77653-922 77640-974 77649-000
1-(2-Fluoro-4-iodo-3-methylphenyl)ethanone ≥95%
1-(2-Fluoro-4-iodo-3-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: C₉H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD34855100
CAS Number: 2386605-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR