2',6'-Difluoro-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%

Supplier: AOBChem USA

Warning

176902-5G 176902-1G 176902-500MG
77627-940EA 1484.23 USD
77627-940 77640-972 77648-998
2',6'-Difluoro-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%
2',6'-Difluoro-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol

Precaución: For Research Use Only

Formula: 210.2198 g/mol
MW: C₁₂H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD35166837
CAS Number: 1592467-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR