1-(2-Benzothiazolyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

176560-5G 176560-1G 176560-500MG 176560-25G
77631-846EA 1034 USD
77631-846 77636-980 77652-426 77657-752
1-(2-Benzothiazolyl)-1-propanone ≥95%
1-(2-Benzothiazolyl)-1-propanone

Precaución: For Research Use Only

Formula: 191.25 g/mol
MW: C₁₀H₉NOS
Storage Temperature: Ambient
CAS Number: 41881-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR