2-Bromo-6-fluoro-3-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

17631-5G 17631-25G 17631-1G 17631-500MG
77634-212EA 452 USD
77634-212 77657-750 77640-970 77644-966
2-Bromo-6-fluoro-3-isopropoxybenzaldehyde ≥95%
2-Bromo-6-fluoro-3-isopropoxybenzaldehyde

Precaución: For Research Use Only

Formula: 261.088 g/mol
MW: C₁₀H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD33403702
CAS Number: 2624417-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR