5-Bromo-2-methoxy-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

17607-5G 17607-1G 17607-25G 17607-500MG
77631-844EA 648 USD
77631-844 77636-978 77653-918 77648-992
5-Bromo-2-methoxy-4-(methylthio)pyridine ≥97%
5-Bromo-2-methoxy-4-(methylthio)pyridine

Precaución: For Research Use Only

Formula: C₇H₈BrNOS
MW: 234.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543646
CAS Number: 474824-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR