3,4-Dibromo-α-methyl-2-thiophenemethanol ≥95%

Supplier: AOBChem USA

Warning

175878-5G 175878-25G 175878-1G 175878-500MG
77631-842EA 915 USD
77631-842 77653-914 77644-962 77644-964
3,4-Dibromo-α-methyl-2-thiophenemethanol ≥95%
3,4-Dibromo-α-methyl-2-thiophenemethanol

Precaución: For Research Use Only

Formula: C₆H₆Br₂OS
Storage Temperature: Ambient
MDL Number: MFCD22385510
CAS Number: 1394021-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR