1-(3,4-Dibromo-2-thienyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

175852-5G 175852-1G 175852-25G 175852-500MG
77627-938EA 1547.06 USD
77627-938 77636-976 77653-912 77644-960
1-(3,4-Dibromo-2-thienyl)-1-propanone ≥95%
1-(3,4-Dibromo-2-thienyl)-1-propanone

Precaución: For Research Use Only

Formula: C₇H₆Br₂OS
Storage Temperature: Ambient
MDL Number: MFCD21960508
CAS Number: 1510709-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR