4,5-Dimethyl-3-nitro-acetanilide ≥95%

Supplier: AOBChem USA

Warning

17582-5G 17582-1G 17582-25G 17582-500MG
77634-206EA 520 USD
77634-206 77636-972 77653-908 77644-958
4,5-Dimethyl-3-nitro-acetanilide ≥95%
4,5-Dimethyl-3-nitro-acetanilide

Precaución: For Research Use Only

Formula: 208.214 g/mol
MW: C₁₀H₁₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09027011
CAS Number: 857571-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR