2-Chloro-3-fluoro-α,α-dimethyl-4-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

175434-5G 175434-500MG 175434-25G 175434-1G
77631-834EA 1366 USD
77631-834 77652-422 77657-744 77640-958
2-Chloro-3-fluoro-α,α-dimethyl-4-pyridinemethanol ≥95%
2-Chloro-3-fluoro-α,α-dimethyl-4-pyridinemethanol

Precaución: For Research Use Only

Formula: 189.61 g/mol
MW: C₈H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD22041922
CAS Number: 40273-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR