6-Methoxy-α,α-dimethyl-3-pyridinemethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
175405-5G
175405-25G
175405-1G
175405-500MG
77631-832EA
1320
USD
77631-832
77653-900
77640-956
77648-982
6-Methoxy-α,α-dimethyl-3-pyridinemethanol ≥95%
6-Methoxy-α,α-dimethyl-3-pyridinemethanol
Precaución: For Research Use Only
Formula:
C₉H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28357366 CAS Number: 1373693-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...