2-Bromo-4-chloro-3-pyridinol ≥97%

Supplier: AOBChem USA

Warning

174675-5G 174675-1G 174675-500MG
77653-898EA 2389 USD
77653-898 77640-952 77644-954
2-Bromo-4-chloro-3-pyridinol ≥97%
2-Bromo-4-chloro-3-pyridinol

Precaución: For Research Use Only

Formula: C₅H₃BrClNO
Storage Temperature: Ambient
MDL Number: MFCD18257594
CAS Number: 1211583-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR