3-Bromo-2-iodo-5-methylthiophene ≥97%

Supplier: AOBChem USA

Warning

174643-5G 174643-1G 174643-25G 174643-500MG
77627-934EA 1516.16 USD
77627-934 77636-968 77653-896 77644-952
3-Bromo-2-iodo-5-methylthiophene ≥97%
3-Bromo-2-iodo-5-methylthiophene

Precaución: For Research Use Only

Formula: C₅H₄BrIS
Storage Temperature: Ambient
MDL Number: MFCD27937158
CAS Number: 89556-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR