2,3-Dichloro-5-ethylthiophene ≥95%

Supplier: AOBChem USA

Warning

174626-5G 174626-25G 174626-1G 174626-500MG
77627-932EA 1484.23 USD
77627-932 77653-894 77640-950 77644-950
2,3-Dichloro-5-ethylthiophene ≥95%
2,3-Dichloro-5-ethylthiophene

Precaución: For Research Use Only

Formula: C₆H₆Cl₂S
Storage Temperature: Ambient
MDL Number: MFCD34148160
CAS Number: 89380-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR