(4,5-Dichlorothiophen-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

174621-1G 174621-5G 174621-500MG
77636-964EA 700 USD
77636-964 77653-890 77644-948
(4,5-Dichlorothiophen-2-yl)boronic acid ≥97%
(4,5-Dichlorothiophen-2-yl)boronic acid

Precaución: For Research Use Only

Formula: C₄H₃BCl₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD18383059
CAS Number: 1190737-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR