5-Bromo-2,3-dichlorothiophene ≥95%

Supplier: AOBChem USA

Warning

174617-5G 174617-1G 174617-25G 174617-500MG
77631-830EA 867 USD
77631-830 77636-962 77653-888 77648-980
5-Bromo-2,3-dichlorothiophene ≥95%
5-Bromo-2,3-dichlorothiophene

Precaución: For Research Use Only

Formula: 231.93 g/mol
MW: C₄HBrCl₂S
Storage Temperature: Ambient
MDL Number: MFCD23135682
CAS Number: 83663-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR