5-Bromo-4-chlorothiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

174612-5G 174612-25G 174612-1G 174612-500MG
77631-828EA 746 USD
77631-828 77653-886 77640-948 77644-946
5-Bromo-4-chlorothiophene-2-carboxylic acid ≥97%
5-Bromo-4-chlorothiophene-2-carboxylic acid

Precaución: For Research Use Only

Formula: C₅H₂BrClO₂S
MW: 241.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08694669
CAS Number: 123418-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR