4,5-Dichloro-2-thiophenecarboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

174610-1G 174610-5G 174610-500MG
77636-960EA 763 USD
77636-960 77657-742 77644-944
4,5-Dichloro-2-thiophenecarboxaldehyde ≥97%
4,5-Dichloro-2-thiophenecarboxaldehyde

Precaución: For Research Use Only

Formula: C₅H₂Cl₂OS
Storage Temperature: Ambient
MDL Number: MFCD22199998
CAS Number: 67482-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR