5-Methoxy-2-(trifluoromethoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

17387-5G 17387-1G 17387-25G 17387-500MG
77631-826EA 1060 USD
77631-826 77636-958 77657-740 77648-978
5-Methoxy-2-(trifluoromethoxy)phenylboronic acid ≥95%
5-Methoxy-2-(trifluoromethoxy)phenylboronic acid

Precaución: For Research Use Only

Formula: 235.953 g/mol
MW: C₈H₈BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD12025981
CAS Number: 1079402-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR