4-Bromo-2,5-dimethoxypyridine ≥95%

Supplier: AOBChem USA

Warning

173691-5G 173691-1G 173691-25G 173691-500MG
77631-822EA 1071 USD
77631-822 77636-954 77657-738 77644-940
4-Bromo-2,5-dimethoxypyridine ≥95%
4-Bromo-2,5-dimethoxypyridine

Precaución: For Research Use Only

Formula: C₇H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD28969395
CAS Number: 1402666-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR