1,3-Dibromo-2-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

17312-5G 17312-25G 17312-1G 17312-500MG
77631-820EA 983 USD
77631-820 77657-734 77640-946 77648-972
1,3-Dibromo-2-propoxybenzene ≥95%
1,3-Dibromo-2-propoxybenzene

Precaución: For Research Use Only

Formula: C₉H₁₀Br₂O
MW: 293.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29948710
CAS Number: 1050424-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR