4'-Ethylbiphenyl-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Ethyl-[1,1-biphenyl]-4-carboxylic acid

172861-5G 172861-25G
77634-196EA 166 USD
77634-196 77657-732
4'-Ethylbiphenyl-4-carboxylic acid ≥97%
4'-Ethylbiphenyl-4-carboxylic acid

Precaución: For Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010210
CAS Number: 5731-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR