8-Bromo-6-ethylquinoline ≥97%

Supplier: AOBChem USA

Warning

172709-5G 172709-25G 172709-1G 172709-500MG
77627-930EA 1484.23 USD
77627-930 77635-484 77640-944 77644-938
8-Bromo-6-ethylquinoline ≥97%
8-Bromo-6-ethylquinoline

Precaución: For Research Use Only

Formula: C₁₁H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD20127909
CAS Number: 91348-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR